Order Entry
Netherlands
ContactUsLinkComponent
 
Organic reference standard, Anthracene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
  EHERL20520000AL
 :  
Organic reference standard, Anthracene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
  EHERL20520000AL
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    120-12-7
  • Description:
    Anthracene 10 µg/ml in acetonitrile
  • Formula:
    C₁₄H₁₀
  • Description:
    Anthracene