Organic reference standard, Chloroform 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA17739500ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:67-66-3
- Description:Chloroform 100 µg/ml in methanol
- Formula:CHCl₃
- Description:Trichloromethane