Order Entry
Netherlands
ContactUsLinkComponent
Organic reference standard, Dichloromethane 100 µg/ml in methanol, Dr Ehrenstorfer
Organic reference standard, Dichloromethane 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA12424500ME
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, Dichloromethane 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA12424500ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    75-09-2
  • Description:
    Dichloromethane 100 µg/ml in methanol
  • Formula:
    CH₂Cl₂
  • Description:
    Dichloromethane

Specifications

About this item