Order Entry
Netherlands
ContactUsLinkComponent
Herbicide reference standards Hexachloro-1,3-butadiene, Dr Ehrenstorfer
Herbicide reference standards Hexachloro-1,3-butadiene, Dr Ehrenstorfer
Supplier:  EHRENSTORFER
Certificates
Herbicide reference standards Hexachloro-1,3-butadiene, Dr Ehrenstorfer
Supplier:  EHRENSTORFER

Specifications

  • CAS No.:
    87-68-3
  • Formula:
    C₄Cl₆
  • Description:
    Hexachloro-1,3-butadiene

Specifications

Frequently Bought Together