Iridium(III) chloride trihydrate (53 - 56% Ir)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13569-57-8
:
Formula:Cl₃Ir·₃H₂O
MDL Number:MFCD00149750
Melting Pt:763 °C (dec.)
Density:5.3 g/cm³
:352.62 g/mol
Storage Temperature:Ambient
EINECS:233-044-6
Specification Test Results
- Appearance:Green to black Crystals
- Assay:53 to 56 % (Ir)
Specification Test Results
Frequently Bought Together