


Cobalt(II) chloride hexahydrate
Catalog # SERA27200.01
Supplier: Serva
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:237.93 g/mol
Merck Index:13,02462
Boiling Pt:110 °C (1013 hPa)
Melting Pt:56 °C
Density:1.92 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-589-4
UN:3077
ADR:9,III
:GHS08
:GHS07
:GHS09
CAS Number:7791-13-1
Formula:CoCl₂·6H₂O
MDL Number:MFCD00149652