Order Entry
Netherlands
ContactUsLinkComponent
Cobalt(II) chloride hexahydrate
Health
Irritant
Environment
Cobalt(II) chloride hexahydrate
Catalog # ROTHT889.1
Supplier:  Roth Carl
CAS Number:  
Health
Irritant
Environment
Cobalt(II) chloride hexahydrate
Catalog # ROTHT889.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications

About this item

 :
 :237.93 g/mol
Merck Index:13,02462
Boiling Pt:110 °C (1013 hPa)
Melting Pt:56 °C
Density:1.92 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-589-4
UN:3077
ADR:9,III
 :GHS08
 :GHS07
 :GHS09
CAS Number:7791-13-1
Formula:CoCl₂·6H₂O
MDL Number:MFCD00149652