


Zinc sulphate monohydrate ≥98%, purified
Catalog # 29245.262
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Assay:Min. 98 %
- Pb (Lead):Max. 100 ppm
- Fe (Iron):Max. 0.012 %
Specification Test Results
About this item
:
:179.45 g/mol
Merck Index:13,10213
Density:3.2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-793-3
UN:3077
ADR:9,III
:GHS07
:GHS05
:GHS09
CAS Number:7446-19-7
Formula:ZnSO₄·1H₂O
MDL Number:MFCD00149893