


Cadmium(II) sulphate hydrate 98% (by complexometric analysis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15244-35-6
:
Formula:CdO₄S·H₂O
MDL Number:MFCD00151655
:226.49 g/mol
Merck Index:14,1627
Melting Pt:41.5 °C trans pt
Density:3.08
EINECS:233-331-6
UN:2570
ADR:6.1,III
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Related Information
Frequently Bought Together