Order Entry
Netherlands
ContactUsLinkComponent
 
Calcium nitrate tetrahydrate
Catalog # LOHM25743.9025
Supplier:  LOHMANN
CAS Number:  
Calcium nitrate tetrahydrate
Catalog # LOHM25743.9025
Supplier:  LOHMANN
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

About this item

 :
 :236.15 g/mol
Merck Index:13,01687
Melting Pt:∼42 °C
Density:1.86 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-332-1
UN:1454
ADR:5.1,III
 :GHS03
CAS Number:13477-34-4
Formula:Ca(NO₃)₂·4H₂O
MDL Number:MFCD00149604