L(+)-Cysteine hydrochloride monohydrate
Supplier: Serva
Certificates
About this item
CAS Number:7048-04-6
:
Formula:HSCH₂CH(NH₂)COOH·HCl·H₂O
MDL Number:MFCD00065606
:175.64 g/mol
Merck Index:13,02810
Melting Pt:168…170 °C
Storage Temperature:Ambient
EINECS:200-157-7
Frequently Bought Together