Order Entry
Netherlands
ContactUsLinkComponent
Ammonium fluoride 30% in aqueous solution
Toxic
Ammonium fluoride 30% in aqueous solution
Catalog # HACH2305726
Supplier:  Hach
CAS Number:  
Toxic
Ammonium fluoride 30% in aqueous solution
Catalog # HACH2305726
Supplier:  Hach
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

About this item

 :
 :37.04 g/mol
Storage Temperature:Ambient
EINECS:235-185-9
UN:3287
ADR:6.1,III
 :GHS06
CAS Number:12125-01-8
Formula:NH₄F
MDL Number:MFCD00011423