Order Entry
Netherlands
ContactUsLinkComponent
Sulphuric acid 50% (v/v) for determination of dissolved oxygen in water
 
Sulphuric acid 50% (v/v) for determination of dissolved oxygen in water
  160346N
 :  
undefined
 
Sulphuric acid 50% (v/v) for determination of dissolved oxygen in water
  160346N
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

 

 

  • Assay:
    49.75 --> 50.25 % (v/v)
  • Substances reducing KMnO4:
    Max. 1 ppm

 

 

 :
 :98.08 g/mol
 :>100 °C (1013 hPa)
 :1.5…1.8 g/cm³ (20 °C)
 :Ambient
 :231-639-5
 :1830
 :8,II
 :GHS05
 :7664-93-9
 :H₂SO₄
 :MFCD00064589