Order Entry
Netherlands
ContactUsLinkComponent
 
 
Orthophosphoric acid 85% ≥85%, HiPerSolv CHROMANORM® for HPLC
  153154D
 :  
undefined
 
Orthophosphoric acid 85% ≥85%, HiPerSolv CHROMANORM® for HPLC
  153154D
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590

 

 

  • Assay:
    Min. 85.0 %
  • Volatile acids:
    Max. 0.0002 meq/g
  • Fe (Iron):
    Max. 10 ppm
  • Pb (Lead):
    Max. 2 ppm
  • Transmittance (254 nm) (0,1 mol/l):
    Min. 80 %

 

 

 :
 :98 g/mol
 :28 °C
 :1.689 g/cm³ (20 °C)
 :Ambient
 :231-633-2
 :1805
 :8,III
 :GHS05
 :7664-38-2
 :H₃PO₄
 :MFCD00011340