
Zinc sulphate 0.05 mol/l (0.05 N) in aqueous solution
Catalog # PRLSMC309336.1000
Supplier: PROLABO CMP
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:161.45 g/mol
EINECS:231-793-3
:GHS09
Storage Temperature:Ambient
CAS Number:7733-02-0
Formula:O₄SZn
MDL Number:MFCD00011302