Order Entry
Netherlands
ContactUsLinkComponent
VWR® IPASEPT® 70, Sterile
VWR® IPASEPT® 70, Sterile
Catalog # 94006.1016
Supplier:  VWR Chemicals
undefined
VWR® IPASEPT® 70, Sterile
Catalog # 94006.1016
Supplier:  VWR Chemicals
Specifications
  • Description:
    IPASEPT® 70, sterile
  • Packed:
    6×1 l trigger spray bottles
  • Pk:
    6 L
Specifications
About this item
Iso-propyl alcohol Ph. Eur. 70% V/V, with purified water Ph. Eur. 30% V/V, filtered at 0,2 μm.
  • Sterilisation by gamma irradiation
  • Sterility Assurance Level (SAL) 10⁻⁶
  • Filled under GMP conditions in cleanroom ISO Class 7
  • Certificate of Analysis available for each batch
  • Fine stream for precise applications, or wide spray for covering large areas
For surface spray cleaning, with or without wipes. Also for cleaning gloved hands before entry into cleanrooms. Ideal for wiping down entrances to controlled environments.