



Glutaraldehyde 8% in aqueous solution
Supplier: Serva
Certificates
About this item
CAS Number:111-30-8
:
Formula:C₅H₈O₂
MDL Number:MFCD00007025
:100.12 g/mol
Storage Temperature:Refrigerator
EINECS:203-856-5
UN:2927
ADR:6.1,II
:GHS06
:GHS05
:GHS08
:GHS09
Frequently Bought Together