Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Inloggen
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Bestelling invoeren
Status van de bestelling
Uw account
Winkelwagen
Netherlands
Bestelling invoeren
Status van de bestelling
Uw account
Winkelwagen
ContactUsLinkComponent
Home
(±)-2-Hexanol
(±)-2-Hexanol
(±)-2-Hexanol
(±)-2-Hexanol
Catalog # EHERC14196800
Supplier:
EHRENSTORFER
CAS Number:
Expand image
(±)-2-Hexanol
Catalog # EHERC14196800
Supplier:
EHRENSTORFER
CAS Number:
Specifications
Seq:
2070
Specifications
About this item
Product Details
:
102.18 g/mol
Storage Temperature:
Ambient
Formula:
C₆H₁₄O
MDL Number:
MFCD00004585
Product Details