Order Entry
Netherlands
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, 1-Methoxy-2-propanol
  65692-5ML-F
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, 1-Methoxy-2-propanol
  65692-5ML-F
 :  Merck
 :  

 

  • Pk:
    5 mL
  • Standards:
    Analytical standard
  • CAS No.:
    107-98-2
  • Description:
    1-Methoxy-2-propanol
  • Seq:
    02
  • Storage conditions:
    RT
  • Hazard class description:
    H226 - H336
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00004537
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Propylene glycol methyl ether; Propyleneglycol monomethyl ether
  • Formula:
    CH₃CH(OH)CH₂OCH₃
  • Molecular weight:
    90,12
  • Density:
    0,916 g/ml at 25 °C (lit.)
  • Flash point:
    82 °C - Closed cup
  • Boiling point:
    118 - 119 °C (lit.)

 

 

  • GHS signal: GHS02, GHS07
 : P210 - P233 - P240 - P241 - P242 - P243