Order Entry
Netherlands
ContactUsLinkComponent
Chloroform ≥99.8% stabilised, Reagent Grade
Chloroform ≥99.8% stabilised, Reagent Grade
Catalog # HONC132950-7L-RC
CAS Number:  
Chloroform ≥99.8% stabilised, Reagent Grade
Catalog # HONC132950-7L-RC
CAS Number:  

Specifications

  • Pack type:
    Stainless steel can, returnable
  • Pk:
    7 L
  • Seq:
    5202

Specifications

Specification Test Results

  • Appearance of the substance:
    complying
  • Purity excluding stabilizer (GC):
    Min. 99.80 %
  • Water (Karl Fischer):
    Max. 0.03 %
  • Ethanol (GC):
    0.5 - 1.0 Gew.%
  • APHA:
    Max. 10
  • IR spectrum:
    complying
  • Non-volatilematter:
    max.0.001%
  • Water (KarlFischer):
    max.0.05%
  • Acid.oralk.react.impurities:
    complying
  • Reaction against H₂SO₄:
    complying
  • Chloride (Cl):
    max.0.0001%
  • Freechlorine:
    complying
  • Foreignchlorinecompounds:
    complying
  • Aldehydes:
    complying
  • Related substances (GC):
    complying

Specification Test Results

About this item

 :
 :119.38 g/mol
Merck Index:13,02160
Boiling Pt:61 °C (1.013 hPa)
Melting Pt:-61.3 °C
Melting Pt:–61.3 °C
Density:1.485 g/cm3 (20 °C)
Storage Temperature:Ambient
EINECS:200-663-8
UN:1888
ADR:6.1,III
 :Stabilised with ethanol 0.5-1.0%
REACH:05-2114614906-45
 :GHS06
 :GHS08
CAS Number:67-66-3
Formula:CHCl₃
MDL Number:MFCD00000826