Chloroform 99+% stabilised, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:67-66-3
:
Formula:CHCl₃
MDL Number:MFCD00000826
:119.38 g/mol
Merck Index:13,02160
Boiling Pt:61 °C (760 mmHg)
Melting Pt:–63 °C
Density:1.48 g/cm³
Storage Temperature:Ambient
UN:1888
ADR:6.1,III
:Stabilised with amylene
EINECS:200-663-8
REACH:05-2114614906-45
:GHS06
:GHS08
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.1 % (K.F.)
- GC:≥99.0 %
- Color scale:≤10 APHA
- Stabilizer:Approx. 100 ppm Amylene
- Residue after evaporation:≤0.002 %
Specification Test Results
Frequently Bought Together