Order Entry
Netherlands
ContactUsLinkComponent
 
Tween® 80 (Polysorbate) for synthesis, Sigma-Aldrich®
 :  Merck
 
Tween® 80 (Polysorbate) for synthesis, Sigma-Aldrich®
 :  Merck
 
 
 

 

  • Identity :
    Passes test
  • Saponification value:
    45 - 55
  • Hydroxyl vlaue:
    65 - 80
  • Density (d20 °C/4 °C):
    1,060 - 1,090
  • pH:
    5 - 7 (50 g/l, H2O, 20 °C)
  • Ignition Point:
    180 °C
  • Viscosity kinematic:
    300 - 500 mm2/s (25 °C)

 

Frequently Bought Together