
TRITON™ X-114 (Polyethylene glycol tert-octylphenyl ether)
Catalog # SERA37243.01
Supplier: Serva
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
Boiling Pt:∼150 °C (1013 hPa)
Density:1.055 g/cm³ (20 °C)
Flash Pt:>93 °C
Storage Temperature:Ambient
UN:3082
ADR:9,III
:GHS05
CAS Number:9036-19-5
MDL Number:MFCD00128254