


TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)
Catalog # SANTSC-29112
Supplier: Santa Cruz Biotechnology
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
Merck Index:13,06793
Boiling Pt:∼200 °C (1013 hPa)
Melting Pt:6 °C
Density:1.06 g/cm³ (20 °C)
Flash Pt:251 °C (closed cup)
UN:3082
ADR:9,III
:GHS05
:GHS07
:GHS09
Storage Temperature:Refrigerator
CAS Number:9002-93-1
:9036-19-5
MDL Number:MFCD00132505