Polyethylene glycol 8,000 for biotechnology
:25322-68-3
:MFCD00081839
:H(OCH₂CH₂)nOH
:MFCD01779615
:8000 g/mol
:12,07729
:>250 °C
:1.21 g/cm³ (20 °C)
:500-038-2
:GHS07
:Ambient
- DNase:None Detected
- pH (5%, Water) @25 °C:4.5 – 7.5
- RNase:None Detected
- Solidification Point:55 – 64 °C
Frequently Bought Together