Order Entry
Netherlands
ContactUsLinkComponent
 
Polyethylene glycol 4,000, powder, EMPROVE® ESSENTIAL Ph. Eur., SAFC®
Catalog # 8.17006.5000
CAS Number:  
Polyethylene glycol 4,000, powder, EMPROVE® ESSENTIAL Ph. Eur., SAFC®
Catalog # 8.17006.5000
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    5 kg
  • Seq:
    4114

Specifications

Specification Test Results

  • Identity:
    passes test
  • Appearance:
    white or almost white solid with a waxy or paraffin-like appearance
  • Appearance of solution:
    passes test
  • Acidity,Alkalinity:
    passes test
  • Reducing substances:
    passes test
  • Density (d 20/20):
    about 1.080 (50% solution)
  • Solidification point:
    53 - 59 °C
  • Viscosity (20 °C):
    106 - 131 mm²/s
  • Viskosity (20 °C):
    114 - 142 mPa∙s
  • Formaldehyde:
    ≤ 30 ppm
  • Ethylene oxide (GC):
    ≤ 1 ppm
  • Dioxane (GC):
    ≤ 10 ppm
  • Hydroxyl value:
    25 - 32
  • Average molecular mass:
    3700 - 4400
  • Sulfated ash:
    ≤ 0.2 %
  • Water (K. F.):
    ≤ 1.0 %

Specification Test Results

About this item

 :
 :4000 g/mol
Boiling Pt:∼250 °C (1013 hPa)
Melting Pt:53…58 °C
Density:1.13 g/cm³ (20 °C)
Flash Pt:250 °C
Storage Temperature:Ambient
EINECS:500-038-2
CAS Number:25322-68-3
 :MFCD00081839
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD01779612