Order Entry
Netherlands
ContactUsLinkComponent
Human albumin (from serum)
Human albumin (from serum)
Supplier:  Serva
Certificates
Human albumin (from serum)
Supplier:  Serva

Specifications

  • CAS No.:
    70024-90-7
  • Source:
    Serum
  • Species:
    Human
  • Storage conditions:
    Refrigerator
  • Protein synonyms:
    Human Serum Albumin|HSA|serum albumin|HAS|serum of albumin
  • Protein/peptide name:
    Albumin

Specifications

Frequently Bought Together