Order Entry
Netherlands
ContactUsLinkComponent
DL-Sodium lactate 50% in aqueous solution
DL-Sodium lactate 50% in aqueous solution
Catalogus # SERA29767.01
Supplier:  Serva
CAS Number:  
DL-Sodium lactate 50% in aqueous solution
Catalogus # SERA29767.01
Supplier:  Serva
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 ml
  • Seq:
    2275

Specifications

About this item

 :
 :112.06 g/mol
Density:1.27…1.32 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-772-0
CAS Number:72-17-3
 :312-85-6
Formula:H₃CCH(OH)COONa
MDL Number:MFCD00065400