Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Starch, soluble

Starch, soluble

Glass bottle for solids, 500 g
Catalog # DIFC217820
Marketsource
Supplier:   DIFCO BD
Starch, soluble
Catalog # DIFC217820
Supplier:  DIFCO BD
Marketsource
CAS Number:  9005-84-9
Starch, soluble

Glass bottle for solids, 500 g

CAS Number:  9005-84-9
Price
153,55 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Starch, Vitex® iodine indicator, Amylodextrin
  • CAS Number:9005-84-9
  • Formula:(C₆H₁₀O₅)n
  • MDL Number:MFCD00082026
  • Molecular Weight:162.14 g/mol
  • Merck Index:13,08878
  • Storage Temperature:Ambient
  • EINECS:232-686-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.