

Streptomycin sulphate
Catalog # PROOICRS0416
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:1457.4 g/mol
Merck Index:12,08983
Melting Pt:250 °C
Storage Temperature:Refrigerator
EINECS:223-286-0
UN:2811
ADR:6.1,III
:GHS08
:GHS07
CAS Number:3810-74-0
Formula:C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
MDL Number:MFCD00037023