Order Entry
Netherlands
ContactUsLinkComponent
Deuterium bromide 48% in D₂O (99% D) (Isotopic)
Deuterium bromide 48% in D₂O (99% D) (Isotopic)
Catalog # 8.15015.0010
Supplier:  Merck
CAS Number:  
Deuterium bromide 48% in D₂O (99% D) (Isotopic)
Catalog # 8.15015.0010
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • Seq:
    2275

Specifications

About this item

 :
 :81.9
Storage Temperature:Ambient
EINECS:236-894-6
CAS Number:13536-59-9
Formula:DBr
MDL Number:MFCD00044232