

Fullerene C60 ≥99.9%, powder, sublimated
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:99685-96-8
:
Formula:C₆₀
MDL Number:MFCD00151408
Merck Index:13,01447
Boiling Pt:500…600 °C (subl.)
Melting Pt:> 280 °C
Density:1.7
UN:1325
ADR:4.1,III
:720.66 g/mol
:GHS02
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together