
Paraffin, highly liquid Ph. Eur., BP, NF, JP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:8002-74-2
Boiling Pt:300 - 450 °C (1013 hPa)
Density:0.85 g/cm³ (20 °C)
Flash Pt:>180 °C
EINECS:232-455-8
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Identity:passes test
- Purity:passes test
- Acidity or alkalinity:passes test
- Density (d 20 °C/20 °C):0.830 - 0.870
- Dynamic viscosity (20 °C):25.0 - 80.0 mPa∙s
- Kinematic Viscosity (40 °C):≤ 33.5 mm²/s
- Arsenic (As):≤ 2 ppm
- Heavy metals (as Pb):≤ 10 ppm
- Residual solvents (Ph.Eur./ICH):excluded by manufactoring process
- Aromatic, polycyclic Hydrocarbons:passes test
- Solid Paraffines:passes test
- Readily carbonisable substance:passes test
- Sulphur compounds:passes test
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERP1.07174.1000_TDS_EN MERP1.07174.2500_TDS_EN MERP1.07174.9026_TDS_EN ProductPage_107174
Product specification
msds_107174
SDS
Frequently Bought Together