Order Entry
Netherlands
ContactUsLinkComponent
Cobalt ≥99.995% (metals basis), foil, Puratronic®, Thickness: 0.5 mm (0.02 in)
Health
Cobalt ≥99.995% (metals basis), foil, Puratronic®, Thickness: 0.5 mm (0.02 in)
Catalog # 11451.FY
CAS Number:  
Health
Cobalt ≥99.995% (metals basis), foil, Puratronic®, Thickness: 0.5 mm (0.02 in)
Catalog # 11451.FY
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001
  • Pk Info:
    50×100 mm

Specifications

About this item

 :
 :58.93 g/mol
Merck Index:12,02488
Boiling Pt:2870 °C (1013 hPa)
Melting Pt:1495 °C
Density:8.9 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-158-0
 :GHS08
CAS Number:7440-48-4
Formula:Co
MDL Number:MFCD00010935