Tungsten ≥99.95% (metals basis), wire, Ø 0.25 mm (0.01 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-33-7
:
Formula:W
MDL Number:MFCD00011461
:183.84 g/mol
Merck Index:12,09945
Boiling Pt:5660 °C (1013 hPa)
Melting Pt:3410 °C
Density:19 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-143-9
Frequently Bought Together