
Niobium ≥99.8% (metals basis), powder APS 1-5 µm
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-03-1
:
Formula:Nb
MDL Number:MFCD00011126
:92.91 g/mol
Storage Temperature:Ambient
EINECS:231-113-5
UN:3089
ADR:4.1,II
:GHS02
Related Information
Frequently Bought Together