Hafnium ≥99.5% (metals basis excluding Zr), foil (Zr ∼ 3%), annealed, Thickness: 1.0 mm (0.04 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-58-6
:
Formula:Hf
MDL Number:MFCD00011032
:178.49 g/mol
Storage Temperature:Ambient
EINECS:231-166-4
Frequently Bought Together