Order Entry
Netherlands
ContactUsLinkComponent
Scandium 15.9 mm (0.63 in) Ø
Scandium 15.9 mm (0.63 in) Ø
Catalog # 45118.KF
CAS Number:  
Scandium 15.9 mm (0.63 in) Ø
Catalog # 45118.KF
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

CAS Number:7440-20-2
 :
Formula:Sc
MDL Number:MFCD00016323
 :44.96 g/mol
Merck Index:12,08537
Boiling Pt:2730 °C (1013 hPa)
Melting Pt:1541 °C
Density:2.989 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-129-2