Order Entry
Netherlands
ContactUsLinkComponent
Gadolinium ≥99% (REO, rare earth oxide basis), ingot/button, Ø ∼50.8 mm (2.0 in)
Gadolinium ≥99% (REO, rare earth oxide basis), ingot/button, Ø ∼50.8 mm (2.0 in)
Catalog # 45106.KF
CAS Number:  
Gadolinium ≥99% (REO, rare earth oxide basis), ingot/button, Ø ∼50.8 mm (2.0 in)
Catalog # 45106.KF
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

CAS Number:7440-54-2
 :
Formula:Gd
MDL Number:MFCD00011022
 :157.25 g/mol
Storage Temperature:Ambient
EINECS:231-162-2