Aluminium ≥99.99% (metals basis), shot, flattened/spherical shape ∼ 9.5 mm (0.375 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7429-90-5
:
Formula:Al
MDL Number:MFCD00134029
:26.98 g/mol
Merck Index:13,00321
Boiling Pt:2327 °C (1013 hPa)
Melting Pt:660 °C
Density:2.7 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-072-3
Frequently Bought Together