
Magnesium ≥99.9% (metals basis), foil, hard, Thickness: 0.05 mm (0.002 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7439-95-4
:
Formula:Mg
MDL Number:MFCD00085308
:24.3 g/mol
Storage Temperature:Ambient
EINECS:231-104-6
UN:1869
ADR:4.1,III
:GHS02
Frequently Bought Together