Bestelling invoeren
Netherlands
ContactUsLinkComponent
Silicone fluid, 100 cSt DC 200
Silicone fluid, 100 cSt DC 200
Catalog # SERA35135.01
Supplier:  Serva
CAS Number:  
Silicone fluid, 100 cSt DC 200
Catalog # SERA35135.01
Supplier:  Serva
CAS Number:  

Specifications

  • Verpakking:
    Glass bottle for solids
  • VPE:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
CAS Number:63148-62-9
 :9016-00-6
MDL Number:MFCD00084411