Nutlin-3 ≥98% (by HPLC)
Catalog #
Product Details & Documents
Inhibitor of p53/MDM2 interaction
(+/-) -Nutlin-3 is an inhibitor of p53/MDM2 interaction (IC50=0.09µM).
Synonyms: 4-(cis-4,5-Bis(4-chlorophenyl)-2-(2-isopropoxy-4-methoxyphenyl)-4,5-dihydro-1H-imidazole-1-carbonyl)piperazin-2-one, (±)-4-[4,5-Bis(4-chlorophenyl)-2-(2-isopropoxy-4-methoxy-phenyl)-4,5-dihydro-imidazole-1-carbonyl]-piperazin-2-once
- CAS Number:548472-68-0
- Formula:C₃₀H₃₀Cl₂N₄O₄
- MDL Number:MFCD07784509
- Molecular Weight:581.49 g/mol



