Amastatin hydrochloride
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Aminopeptidase inhibitor.
Competitive inhibitor of human serum aminopeptidase A, pig kidney leucine aminopeptidase, leucine aminopeptidase, tyrosine aminopeptidase, but not of aminopeptidase B. Has been used to potentiate the action of some bioactive peptides by inhibiting their degradation by aminopeptidases.
Specifications
- MDL No.:MFCD00150636
- Protein/peptide name:Amastatin hydrochloride
- Purity:98%
- Molecular weight:511,013 g/mol
- Molecular formula:C₂₁H₃₉ClN₄O₈
Specifications
Frequently Bought Together