Order Entry
Netherlands
ContactUsLinkComponent
Human recombinant Shp-1 (from E. coli)
Human recombinant Shp-1 (from E. coli)
Catalogus # ENZOBMLSE3340020
Supplier:  Enzo Life Sciences
CAS Number:  
Human recombinant Shp-1 (from E. coli)
Catalogus # ENZOBMLSE3340020
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    20 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Protein/peptide name:
    Shp-1
  • Formulation:
    Liquid. In 50 mM TRIS/HCl, pH 8,0, containing 150 mM NaCl, 5 mM DTT, 0,03% Brij 35, 0,1 mM EDTA and 10% glycerol.

Specifications

About this item

Produced in E. coli. Full length human Shp-1 (aa 1-595).