Order Entry
Netherlands
ContactUsLinkComponent
Human recombinant MMP-12 (catalytic domain) (from E. coli)
Human recombinant MMP-12 (catalytic domain) (from E. coli)
Catalog # ENZOBMLSE1380010
Supplier:  Enzo Life Sciences
CAS Number:  
Human recombinant MMP-12 (catalytic domain) (from E. coli)
Catalog # ENZOBMLSE1380010
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Protein/peptide name:
    MMP-12 (catalytic domain)

Specifications

About this item

Produced in E. coli. Active Matrix Metalloproteinase-12 (MMP-12, metalloelastase, macrophage elastase; often confused with neutrophil elastase) catalytic domain from human cDNA.

The enzyme consists of the catalytic domain of human MMP-12 (Phe99-Leu271, NM_002426) with a C-terminal purification tag.