Order Entry
Netherlands
ContactUsLinkComponent
PDKtide, biotin
PDKtide, biotin
  ENZOBMLP2510100
 :  
PDKtide, biotin
  ENZOBMLP2510100
 :  

 

  • Pk:
    100 µG
  • Conjugation:
    Biotin
  • Seq:
    02
  • Protein/peptide name:
    PDKtide
  • Purity:
    95%

 

 

Protein kinase substrate

This peptide, Biotin-Ahx-KTFCGTPEYLAPEVRREPRILSEEEQEMFRDFDYIADWC, is derived from AKT1 and PKN2/PRK2. It is a substrate for PDK1, CDK9/Cyclin T1 and JAK2.The biotin allows peptide to be used in kinase assays with streptavidin-bound membranes.