Axltide, biotin
ENZOBMLP2490001
:
- Pk:1 mg
- Conjugation:Biotin
- Seq:02
- Protein/peptide name:Axltide
- Purity:95%
Protein kinase substrate
This peptide, biotin-Ahx-KKSRGDYMTMQIG, is derived from mouse insulin receptor 1 and is a substrate for Axl, DDR2, Mst1, and JAK2 kinases. The biotin allows peptide to be used in kinase assays with streptavidin-bound membranes.