Order Entry
Netherlands
ContactUsLinkComponent
Bandeiraea simplicifolia isolectin B4, FITC (fluorescein isothiocyanate)
Bandeiraea simplicifolia isolectin B4, FITC (fluorescein isothiocyanate)
Catalog # ENZOALX650001FMC05
Supplier:  Enzo Life Sciences
CAS Number:  
Bandeiraea simplicifolia isolectin B4, FITC (fluorescein isothiocyanate)
Catalog # ENZOALX650001FMC05
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    0,5 mg
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Species:
    Bandeiraea simplicifolia
  • Seq:
    02
  • Protein/peptide name:
    Isolectin B4
  • Concentration:
    1 mg/ml
  • Formulation:
    Liquid. In 10 mM HEPES, pH 7,5, containing 0,15 M sodium chloride, 0,08% sodium azide and 0,1 mM Ca²⁺.

Specifications

About this item

Isolated from Griffonia (Bandeiraea) simplicifolia seeds.

Binds to alpha-(1,3) galactosyl galactoside epitopes.