Human recombinant IL33 (from E. coli)
Supplier: Enzo Life Sciences
Certificates
About this item
Produced in E. coli. Untagged human IL-33 (aa 112-270).
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Biological activity:Activates human ST2-dependent NF-κB pathway. Does not activate mouse ST2-dependent NF-κB.
- Reconstitution instructions:Reconstitute with 100 µl sterile distilled water. Further dilutions should be made with medium containing 5% fetal calf serum.
- Protein/peptide name:IL33
- Purity:>95% (SDS-PAGE)
- Concentration:0,1 mg/ml after reconstitution
- Formulation:Lyophilised. Contains PBS.
Specifications
Frequently Bought Together